Learn More
Greiner Bio-One ComboPlate™ CrystalBridge Protein Crystallization Plate
Ideal for protein crystallization. Greiner Bio-One ComboPlate™ CrystalBridge Protein Crystallization Plate is used for sitting drop applications.
Brand:  Greiner Bio-One 662145
Product Code. 10315202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Virgin, high-clarity polystyrene combined with distortion-free bottom provides optical properties for microscopic inspection of crystal growth
  • Flattened raised ring around each well reduces risk of cross-contamination
  • Raised protection lid protects cover slips and sealer during transportation and storage
  • Conical well bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Non-sterile
Well: 4.6 mm
Polystyrene
250/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One ComboPlate™ CrystalBridge Protein Crystallization Plate > Protein Crystallization Plate