Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick
Shop All Fisherbrand Products
Click to view available options
Volume (Metric) Well:
40 μL
50 μL
70 μL
80 μL
100 μL
120 μL
200 μL
1300 μL
Unit Size:
Each
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations |
| For Use With (Equipment) | OmniPAGE Mini Vertical Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 20 |
| Quantity | 1 Each |
| Thickness (Metric) | 2 mm |
| Volume (Metric) Well | 40 μL |