Learn More
Fisherbrand™ Combs for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
Product Code. 11716175
Click to view available options
No. of Wells:
8
12
15
20
Volume (Metric) Sample:
10 μL
15 μL
20 μL
25 μL
35 μL
45 μL
50 μL
65 μL
90 μL
Width (Metric) Teeth:
2.5 mm
4 mm
5.5 mm
9.25 mm
Unit Size:
Each
Product Code. 11716175
Brand: Fisherbrand™ TVGC18
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit
  • Available in 1, 1.5 and 2mm thickness
Specifications
For Use With (Equipment) TVG-SYS Vari-Gel MIDI Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 8
Thickness (Metric) 1 mm
Volume (Metric) Sample 45 μL
Width (Metric) Teeth 9.25 mm