Learn More
Cytiva Complete Tubing Nipple
Shop All Cytiva Products
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Specifications
Specifications
| Material | Polypropylene, Glass Fiber Reinforced, Acetal Plastic, Fluoro Rubber, Fluor Carbon Rubber (FPM) |
| Type | Complete Tubing Nipple |
| Includes | 1 x Connector, 1 x Sealing plug, 1 x Connector nμt |
| Quantity | 1 Each |
| Weight (Metric) | 0.1 kg |