Learn More
Conductivity Calibration Solution, Cole-Parmer™ Product Code.: 11709759
Conductivity Calibration Solution, Cole-Parmer™
Product Code. 11709759 Shop All Cole-Parmer Products
20 mL
Click to view available options
Quantity:
20 mL
Product Code. 11709759
Brand: Cole Parmer 3565309
Please to purchase this item. Need a web account? Register with us today!
Specifications
Quantity 20 mL
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    17
    Hours
    10
    Minutes
    41
    Seconds
    32
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    48
    Hours
    10
    Minutes
    41
    Seconds
    32
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Conductivity Calibration Solution, Cole-Parmer™ >