Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
Brand:  JG Finneran Associates™ 4025PF-631
 View more versions of this product
Product Code. 10045510
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Available preassembled with a polymer bottom spring or patented Top Spring™
  • Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
  • Conical design permits complete sample removal
Specifications
Specifications
Polypropylene
Plastic Flange for Step Vial
100 Pk.
250 μL
Big mouth, limited volume, precision formed interior