Learn More
JG Finneran Associates™ Conical Inserts
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
Material:
Glass
Polypropylene
Unit Size:
Pack of 100
Description
- Available preassembled with a polymer bottom spring or patented Top Spring™
- Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
- Conical design permits complete sample removal
Specifications
Specifications
| Material | Glass |
| Volume (Metric) | 100 μL |
| Includes | Top Spring |
| Style | Limited volume, precision formed mandrel |
| Quantity | 100 Pk. |