Learn More
Bollé Safety Contour Platinum™ Smoke Safety Glasses
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Smoke |
Product Type | Contour CONTPSF Safety Spectacle |
No. per Pack | 1 Ea. |
Weight (Metric) | 21 g |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Videos
-
Bolle Safety Platinum efficiency demonstration
-
Bolle Safety Platinum coating
Documents
Product Information
Specification Sheets
CONTOUR
File size: 752.3 kB
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bollé Safety Contour Platinum™ Smoke Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction