missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, ≥98.0%, Reagent Grade, Honeywell™

Product Code. p-7120356 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 15687770

Brand: Honeywell Research Chemicals C7631250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper;sulfate;pentahydrate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 110°C
Color Blue Green
pH 3.5 to 4.5 (at 20°C)
Physical Form Solid
Quantity 250 g
Linear Formula CuSO4 · 5H2O
UN Number UN3077
Formula Weight 249.69g/mol
Percent Purity ≥98.0%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 7.3mmHg (25°C)
Density 2.28g/cu.cm at 20°C
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.