missing translation for 'onlineSavingsMsg'
Learn More

Copper phthalocyanine, dye content ca 95%

Product Code. 10489940 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
This item is not returnable. View return policy

Product Code. 10489940

Brand: Thermo Scientific Acros 269152500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 147-14-8 | C32H16CuN8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160, Pigment blue 15

Specifications

Merck Index 15, 2505
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 98% h2so4, practically insoluble in alcohol and hydrocarbons
Infrared Spectrum Authentic
Formula Weight 576.08
Percent Purity ≥94% (ex Cu)
Packaging Glass bottle
UV Spectrometry Complies
Color Blue
Quantity 250 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Copper phthalocyanine, dye content ca 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.