Learn More
Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
Brand:  Corning™ 91-200-41
 View more versions of this product
Product Code. 17273574
  • 55.30 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.5 L
EVA
1/Pk.
Each
Heat sealable flexible tubing
1/Ea.
Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Collection Bags > EVA; 500 mL