Learn More
Corning™ Smooth 1536-Well Multiple Well Plates
GREENER_CHOICE
Product Code. 12761935 Shop All Corning Products
Click to view available options
Color:
Black
White
Unit Size:
Pack of 50
Product Code. 12761935
Brand: Corning™ 7247
Only null left
Add to basket
Add to basket
Ensure precision liquid handling
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 32 x 48
Color White
Product Type 1536-Well Assay Microplate
For Use With (Equipment) Automated dispensers including the PerkinElmer FlexDrop™
Material Polystyrene
Volume (Metric) Well 12.5 μL
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For Kalypsys™ systems, ensure precision liquid handling
No. per Case 50/Cs.
No. per Pack 10/Pk.