Learn More
Falcon™ Vented Screw Caps for Flasks, Sterile
Designed for use with Falcon 25-, 75-, and 175cm2 flasks
Brand: Falcon™ 354639
This item is not returnable.
View return policy
Description
- Ensure consistent gas exchange and minimize contamination
- Polyethylene caps with an integral, hydrophobic 0.2 μm microporous membrane filter vent allow consistent gas exchange when the caps are in the closed position.
- Gases required or cell growth and metabolism pass freely through the vent while microorganisms cannot. Falcon vented caps will not wet out.
- Vented caps minimize contamination associated with standard open incubation. The vented caps prevent media that can become trapped in a partially opened cap from blocking gas exchange. The caps will not fall off in the incubator because of vibration.
Specifications
Polyethylene | |
Falcon™ 175 cm2 flasks | |
Cap, Vented | |
Screw Cap |
Blue | |
0.2μm Membrane vent | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Falcon™ Vented Screw Caps for Flasks, Sterile > 175 cm2
Spot an opportunity for improvement?Share a Content Correction