Learn More
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
Brand: Corning™ 8160
272.95 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
- Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Specifications
Cellulose Acetate | |
Sterile | |
0.5 mL |
0.22 μm | |
Spin-X |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
36Hours
2Minutes
34Seconds
12New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.