Learn More
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Product Code. 10787632 Shop All Corning Products
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 96
Product Code. 10787632
Brand: Corning™ 8162
Please to purchase this item. Need a web account? Register with us today!
Edge
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Cellulose Acetate
Pore Size 0.45 μm
Sterility Sterile
Type Spin-X
Volume (Metric) 0.5 mL