Learn More
Corning™ Costar™ Netwell™ Inserts
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers.
Marca: Corning™ 3480
Descripción
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
Especificaciones
2 x 3 | |
Clear | |
Round | |
Cell Culture Insert | |
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers | |
6/Pk. | |
440 μm |
Yes | |
Polystyrene | |
Sterile | |
6 | |
48/Cs. | |
8 inner packs of 6 | |
48/Cs. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.