Learn More
DWK Life Sciences Cover Slips, from D 263™ M
Shop All DWK Life Sciences ProductsDescrizione
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be dyed in a dye bath.
- Pure white borosilicate glass (D 263™ M) is absorption-free in the visible spectral range
- High optical homogeneity of the glass and thus practically free of surface defects, inclusions, streaks, bubbles and annoying streaks due to optimal manufacturing process
- Highly transparent and colourless (colour neutrality)
- High flatness enables optimum working conditions
- Very uniform wettability
Specifica
Specifica
| Material | Borosilicate Glass D 263 M |
| Length (Metric) | 60 mm |
| Width (Metric) | 24 mm |
| Thickness (Metric) | 1.5 mm |
| For Use With (Application) | Covering and fixing of samples during microscopy; Droplet distribution on the slide; Use in microscope slide staining or automatic covering machines |
| Quantity | 10 x 100/Pk. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.