Learn More
Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories
Product Code. p-4530497 Shop All Thermo Scientific Products
Click to view available options
Type:
Cane Holder
Straw Holder
Vial Holder
Unit Size:
Each
Product Code. 10795505
Brand: Thermo Scientific™ 4000710
Please to purchase this item. Need a web account? Register with us today!
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
TRUSTED_SUSTAINABILITY
Specifications
Type Straw Holder
For Use With (Equipment) All CryoMed Controlled-Rate Freezers
Holds Straw