Learn More
Thermo Scientific™ Culti-Loops™ Stenotrophomonas maltophilia ATCC™ 5133™
Product Code. 12995783
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
Product Code. 12995783
Brand: Thermo Scientific™ R4607064
Please to purchase this item. Need a web account? Register with us today!
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 51331TM*
Description Stenotrophomonas maltophilia ATCC™ 51331™
Format Culti-Loops
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-negative Rods
Quantity 5 Loops/Pack
Strain Designations 89-02-019
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.