Learn More
Ambion™ Custom mirVana™ miRNA Mimic
Product Code. p-7075764
Click to view available options
Quantity:
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
For Research Use Only. All usage must comply with product instructions.
Product Code. 15531124
Brand: Ambion™ 4464068
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Custom mirVana™ miRNA Mimic
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. Use our GeneAssist™ miRNA Workflow Builder to design both mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C).
Description mirVana Custom miRNA Mimic, 5nmol, Lowest Off-Target Effects and Highest Potency, miRNA mimics are small, Chemically Modified Double-Stranded RNAs that mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-Regulation of miRNA Activity
RNAi Type miRNA
Product Type mirVana Custom miRNA Mimic
Quantity 5 nmol