Learn More
Ambion™ Custom Select siRNA
Product Code. 15561084
Click to view available options
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
5 nmol
Unit Size:
20nmol
250nmol
40nmol
5nmol
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 15561084
Brand: Ambion™ 4444207
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Custom Select siRNA
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C
RNAi Type siRNA
Product Line Silencer Select, Ambion
Product Type siRNA
Purity HPLC
Quantity 10 μmol