missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
189.00 €
Specifications
| Antigen | CXCR4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 4 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Knockdown Validated |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| CXCR4 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 4 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Cancer, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
| PBS with 0.02% Sodium Azide | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
| CXCR4 | |
| IgG | |
| Peptide affinity purified | |
| Human CXCR4 has four isoforms, including isoform a (356aa, 40.2kD), isoform b (352aa, 39.7kD), isoform c (423aa, 47.6kD), and isoform d (385aa, 43.4kD). This antibody detects human isoform b only, but not other human isoforms. Mouse CXCR4 has two isoforms, including isoform 1 (359aa, 40.4kD) and isoform 2 (357aa, 40.2kD). Rat CXCR4 has only one isoform identified so far (349aa, 39.4kD). |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_003458 | |
| 7852 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the center of human CXCR4 isoform b. The immunogen is located within amino acids 170-220 of CXCR4. Amino Acid Squence: DRYICDRFYPNDLWV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 40 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title