Learn More
Cytiva Cy7 Bis NHS Ester
Suitable for general labeling of amine residues
Brand: Cytiva PA17002
This item is not returnable.
View return policy
Description
The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.- Suitable for general labeling of amine residues.
- Large range of bulk packs available for greatest economy.
Specifications
100% Cy7 (Bis NHS ester) | |
50 mg | |
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. | |
Purple |
Used for high-throughput screening (HTS) applications | |
Cy7 Bis NHS Ester | |
Cy7 |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Specific target organ toxicity Category 2
Hazard Statement
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Cy7 (Bis NHS ester)