Learn More
KARTELL Cylindrical Test Tube
The Cylindrical Test Tube has a tight screw cap for sealed test environments.
Brand: KARTELL 8821000
Description
- Tight screw cap
- Taking into consideration the different applications, they are available in sterile single packed or non-sterile multi-packed.
Specifications
20 mL | |
Polystyrene | |
500 pcs. | |
Not Autoclavable | |
Test Tube |
150 mm | |
Cylindrical | |
Non-sterile | |
No |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.