Learn More
Invitrogen™ CYP2B6 BACULOSOMES™ Plus Reagent, rHuman
Product Code. 15481194
Click to view available options
Quantity:
0.5 nmol
Unit Size:
0.5nmol
For Research Use Only. All usage must comply with product instructions.
Product Code. 15481194
missing translation for 'mfr': Invitrogen™ P3028
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
CYP2B6 BACULOSOMES™ Plus Reagent, rHuman
Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase. For this particular isozyme (CYP2B6), human cytochrome b5 is also included.
  • Single human P450 isozyme for detailed drug metabolism studies
  • High specific activity increases dynamic range and sensitivity
  • No background metabolism for clear results
    Single Overexpressed Human CYP450 Isozyme
    CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over Human Liver Microsomes (HLMs) in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s or other classes of drug metabolizing enzymes.
    High Activity Results in Increased Sensitivity
    Compound rankings based on metabolism and inhibition profiles observed with CYP450 BACULOSOMES™ Plus Reagents are very similar to those seen with HLMs for most compounds tested. However, CYP450 BACULOSOMES™ Plus Reagents activity and metabolic rates with most substrates is significantly higher than those seen with HLMs. This results in a broad dynamic range and high sensitivity in assays utilizing CYP450 BACULOSOMES™ Plus Reagents. The high level of activity and reproducibility of the enzyme component observed in the reaction with most probe substrates makes CYP450 BACULOSOMES™ Plus Reagents well suited to high-throughput screening formats.
    High Signal to Noise Ratio
    No background metabolism by endogenous insect CYP450s has been detected with any of the Vivid™ probe substrates tested so far.
    Applications: isozyme identification, DMPK analysis, isozyme-specific metabolism, and inhibition screening
    For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Line BACULOSOMES
    Content And Storage Store in ultra-cold freezer (-68 to -85°C).
    Product Type Bacterial Microsome
    Quantity 0.5 nmol
    Format Liquid suspension
    Shipping Condition Dry Ice
    Research Category Toxicology & Drug Metabolism