missing translation for 'onlineSavingsMsg'
Learn More

Cytidine-5'-Triphosphate Disodium Salt Dihydrate

Product Code. 15825278 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
Packaging:
Plastic bottle
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 15825278

Brand: Thermo Scientific J14121.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 81012-87-5 | C9H18N3Na2O16P3 | 563.15 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-87-5
Molecular Formula C9H18N3Na2O16P3
Molecular Weight (g/mol) 563.15
MDL Number MFCD00150807
InChI Key KNSYTSUGTLJHPS-ODQFIEKDSA-L
Synonym cytidine 5 '-triphosphate sodium salt, cytidine 5 '-triphosphate tetrasodiμm
PubChem CID 129893565
IUPAC Name [[(2 R,3 S,4 R,5 R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodiμm
SMILES O.O.[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Physical Form Powder
Quantity 100 mg
Formula Weight 527.13 (anhydrous)
Packaging Plastic bottle
Chemical Name or Material Cytidine to 5 '-Triphosphate Disodium Salt Dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.