Learn More
Cytokeratin, pan Antibody (AE-1/AE-3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems NBP2-33200AF532
Product Code. 18615169
  • 527.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit, Reptile, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF, Single Cell Western, Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin, pan
Monoclonal
Alexa Fluor 532
AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK
Human epidermal keratin
0.1 mL
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
3848
Store at 4°C in the dark. Do not freeze.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
AE-1/AE-3
50 mM Sodium borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat, Bovine, Canine
Purified