Learn More
Thermo Scientific™ Cytomat™ 2 C425-LiN Automated Incubator, Copper
Shop All Thermo Scientific ProductsDescription
Ensure a safe, reproducible process, unsurpassed temperature uniformity and stability with the next generation of mid-range Thermo ScientificTM CytomatTM 2 C4xx-LiN series automated incubators for incubation/storage of cell culture applications.
These Cytomat incubators are available in various temperature ranges and gate positions for the hand-off to the lab automation robot. The device can be seamlessly integrated with a mean access time of 12 sec.
With a fully automated thermal decontamination and optional controlled humidity, the Cytomat 2 C4xx-LiN series provides a new standard in significantly reducing the risk of contamination.
The Cytomat 2 C4xx-LiN series of automated incubators provide an ideal safe environment for storage and incubation of assays. This series of mid-range incubators offer a choice of temperature ranges (4 to 70°C), high humidity up to 95% r.H. stable CO2 conditions, and automated ContraCon decontamination routine without harmful gases.
The external cooling and heating unit is designed to allow the user to create a customized working space for easy system integration into any automated assay system.
Available options
- Integrated barcode reader (1D/2D) enables sample tracking, barcode transmission, and inventory management.
- P3-Plate detection in stacker helps to eliminate labware handling errors.
- HydraSmart Technology for precise humidity control without heat extraction and external water tank to avoid contamination.
- Right speed setting for the right application with individual speed setting per plate and adjustable incremental speed settings (Slow/Medium/Fast).
- Flexible positioning inside your lab with 6 possible gate positions including under bench in a single body.
- Solid Copper inner chamber for natural contamination prevention.
- Tower Shaker (ToS) for TRUE orbital shaking of applications requiring sample agitation and cells that need to be kept in suspension.

Specifications
Specifications
Certifications/Compliance | CE Marked |
Access Time | 12 sec. |
Chamber Material | Copper |
Holds | 56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well |
Relative Humidity | <90% |
Temperature Range (Metric) | Ambient +4° to 25°C |
Type | Automated Incubator |
Features | Incubation and storage unit with temperature range above; Humidity up to 90% r.H.; Includes decontamination routine-ContraCon |
For Use With (Application) | Incubation/storage for cell culture applications 4 to 25°C |
Model | Cytomat 2 C425-LiN 230V 50Hz |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.