Learn More
Epredia™ Cytoslide™ Microscope Slides
Improve processing and visualization of small-volume specimens with these cytoslides, for use with Epredia™ Shandon™ EZ Single Cytofunnels™.
Brand: Epredia™ 5991059
47.06 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Uncoated slides available with circle on front or back
- Available in boxes of 100 slides
Specifications
Single Cytoslide | |
Plain | |
Noncoated, circle on front |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
26Hours
19Minutes
29Seconds
26 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.