Learn More
Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99%
Chemical Identifiers
CAS | 5794-24-1 |
---|---|
Molecular Formula | C4H10N2O4 |
Molecular Weight (g/mol) | 150.134 |
MDL Number | MFCD00149558 |
InChI Key | RBMGJIZCEWRQES-HSHFZTNMSA-N |
Synonym | d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide |
PubChem CID | 16211197 |
IUPAC Name | (2R)-2,4-diamino-4-oxobutanoic acid;hydrate |
SMILES | C(C(C(=O)O)N)C(=O)N.O |
Description
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water (35 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
5794-24-1 | |
150.134 | |
RBMGJIZCEWRQES-HSHFZTNMSA-N | |
16211197 | |
C(C(C(=O)O)N)C(=O)N.O |
C4H10N2O4 | |
MFCD00149558 | |
d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide | |
(2R)-2,4-diamino-4-oxobutanoic acid;hydrate |
Specifications
274°C to 276°C (decomposition) | |
C4H10N2O4 | |
14,837 | |
Soluble in water (35g/L at 20°C). | |
C(C(C(=O)O)N)C(=O)N.O | |
150.134 | |
150.14 (132.12 Anhydrous) | |
D-(-)-Asparagine monohydrate |
5794-24-1 | |
MFCD00149558 | |
d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide | |
RBMGJIZCEWRQES-HSHFZTNMSA-N | |
(2R)-2,4-diamino-4-oxobutanoic acid;hydrate | |
16211197 | |
99% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only