missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Aspartic acid, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
D-Aspartic acid modulates melatonin synthesis in the pineal gland.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Aspartic acid modulates melatonin synthesis in the pineal gland.
Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
D-Aspartic acid modulates melatonin synthesis in the pineal gland.
Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | >300°C (decomposition) |
Beilstein | 1723529 |
Merck Index | 14,840 |
Solubility Information | Slightly soluble in water,Soluble in 1N ammonium hydroxide (100mg/ml),and dilute HCl. Insoluble in ether,and alcohol. |
Formula Weight | 133.1 |
Percent Purity | 99% |
Density | 1.66 |
Quantity | 100 g |
Optical Rotation | −25° (c=1 in 1N HCl) |
Chemical Name or Material | D-Aspartic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'