missing translation for 'onlineSavingsMsg'
Learn More

D-Cysteine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

26.20 € - 79.20 €

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11925721
View Documents Promotion Details
Thermo Scientific Acros
442782500
250 mg Glass Bottle
26.20 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11915721
View Documents Promotion Details
Thermo Scientific Acros
442785000
500 mg Glass Bottle
45.20 €
500mg
Please to purchase this item. Need a web account? Register with us today!
11905721
View Documents Promotion Details
Thermo Scientific Acros
442780010
1 g Glass Bottle
79.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207121-46-8
175.627
QIJRTFXNRTXDIP-YBBRRFGFSA-N
10176341
C(C(C(=O)O)N)S.O.Cl
C3H10ClNO3S
MFCD00150051
d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
(2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride

Specifications

207121-46-8
C3H10ClNO3S
d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
C(C(C(=O)O)N)S.O.Cl
175.627
177.77
98%
White to Yellow
MFCD00150051
QIJRTFXNRTXDIP-YBBRRFGFSA-N
(2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
10176341
Crystalline Powder
D-Cysteine hydrochloride monohydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 251-043-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only