missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Cysteine hydrochloride monohydrate, 99%

85.30 € - 283.00 €

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11406200
View Documents Promotion Details
Thermo Scientific Chemicals
H27107.03
1 g
85.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
11416200
View Documents Promotion Details
Thermo Scientific Chemicals
H27107.06
5 g
283.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207121-46-8
175.627
QIJRTFXNRTXDIP-YBBRRFGFSA-N
10176341
C(C(C(=O)O)N)S.O.Cl
C3H10ClNO3S
MFCD00150051
d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
(2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride

Specifications

184°C to 186°C
C3H10ClNO3S
d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
C(C(C(=O)O)N)S.O.Cl
175.627
175.64 (157.62 Anhydrous)
−5.5° (c=5 in 5N HCl)
207121-46-8
MFCD00150051
QIJRTFXNRTXDIP-YBBRRFGFSA-N
(2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
10176341
99%
D-Cysteine hydrochloride monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 251-043-9

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only