missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15437076
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27025-25-8
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.642
MDL Number MFCD00235872
InChI Key MFUPLHQOVIUESQ-NUBCRITNSA-N
Synonym dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride
PubChem CID 12917567
IUPAC Name dimethyl (2R)-2-aminopentanedioate;hydrochloride
SMILES COC(=O)CCC(C(=O)OC)N.Cl

Specifications

Melting Point 91°C
Formula Weight 211.64
Percent Purity 98%
Quantity 25 g
Optical Rotation −26° (c=5 in Water)
Sensitivity Hygroscopic
Chemical Name or Material D-Glutamic acid dimethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'