missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Methionine, 99+%
44.30 € - 418.00 €
Chemical Identifiers
CAS | 348-67-4 |
---|---|
Molecular Formula | C5H11NO2S |
Molecular Weight (g/mol) | 149.21 |
MDL Number | MFCD00002622 |
InChI Key | FFEARJCKVFRZRR-SCSAIBSYSA-N |
Synonym | d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid |
PubChem CID | 84815 |
ChEBI | CHEBI:16867 |
SMILES | CSCC[C@@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10185090
|
Thermo Scientific Chemicals
227210050 |
5 g | Glass Bottle |
44.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10195090
|
Thermo Scientific Chemicals
227210250 |
25 g | Glass Bottle |
132.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15179253
|
Thermo Scientific Chemicals
227211000 |
100 g | Glass Bottle |
418.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
348-67-4 | |
149.21 | |
FFEARJCKVFRZRR-SCSAIBSYSA-N | |
84815 | |
CSCC[C@@H](N)C(O)=O |
C5H11NO2S | |
MFCD00002622 | |
d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
CHEBI:16867 |
Specifications
273.0°C to 275.0°C | |
348-67-4 | |
White | |
10ppm max. | |
C5H11NO2S | |
CH3SCH2CH2CH(NH2)CO2H | |
MFCD00002622 | |
(TLC) not detectable | |
Solubility in water: soluble in water: 53g/L (20°C). | |
CSCC[C@@H](N)C(O)=O | |
− 24.00 | |
84815 | |
300ppm max. | |
98% min. (c=3, H2O, 430nm) 1cm cell | |
0.1% max. | |
Powder |
200ppm max. | |
1ppm max. | |
300ppm max. | |
99+% | |
Authentic | |
10ppm max. | |
0.3% max. | |
d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
FFEARJCKVFRZRR-SCSAIBSYSA-N | |
(2R)-2-amino-4-(methylsulfanyl)butanoic acid | |
149.21 | |
CHEBI:16867 | |
149.21 | |
≥99% | |
− 24.00 (20.00°C c=8,6N HCl) | |
D-Methionine, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 206-483-6
missing translation for 'rtecsNumber' : PD0455000
missing translation for 'tsca' : TSCA
RUO – Research Use Only