Learn More
Dactolisib Tosylate, MedChemExpress
MedChemExpress Dactolisib Tosylate (BEZ235 Tosylate) is a dual PI3K and mTOR kinase inhibitor with IC50 values of 4, 75, 7, 5 nM for PI3Kα, β, γ, δ, respectively. Dactolisib Tosylate (BEZ235 Tosylate) inhibits mTORC1 and mTORC2.
Brand: Medchem Express HY-15174
Description
Dactolisib Tosylate (BEZ235 Tosylate) is a dual PI3K and mTOR kinase inhibitor with IC50 values of 4, 75, 7, 5 nM for PI3Kα, β, γ, δ, respectively. Dactolisib Tosylate (BEZ235 Tosylate) inhibits mTORC1 and mTORC2.Specifications
Dactolisib Tosylate | |
White | |
C37H31N5O4S | |
BEZ235 TosylateNVP-BEZ 235 Tosylate | |
CN(C1=C2C3=CC(C4=CC5=CC=CC=C5N=C4)=CC=C3N=C1)C(N2C6=CC=C(C=C6)C(C)(C#N)C)=O.CC7=CC=C(S(=O)(O)=O)C=C7 | |
641.74 | |
Research | |
Cancer-Kinase/protease |
1028385-32-1 | |
Research | |
100 mg | |
DMSO : 34 mg/mL (52.98 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble) | |
641.74 | |
98.8% | |
Solid |