missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX6 Recombinant Rabbit Monoclonal Antibody (JB41-32)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534652
This item is not returnable.
View return policy
Description
Positive Control: Rat brain tissue, human tonsil tissue, human kidney tissue, mouse small intestine tissue, human colon tissue, SH-SY-5Y, K562, SiHa.
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
Specifications
| DDX6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P26196 | |
| DDX6 | |
| Recombinant protein within Human DDX6 aa 81-220. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JB41-32 | |
| Unconjugated | |
| DDX6 | |
| 1110001P04Rik; ATP-dependent RNA helicase p54; DDX6; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); DEAD-box helicase 6; DEAD-box protein 6; E230023J21Rik; HLR2; mRCK/P54; Oncogene RCK; oncogene RCK homolog; p54; probable ATP-dependent RNA helicase DDX6; RCK; RGD1564560 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1656 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction