missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ deconex™ 53 PLUS Disinfectant

Product Code. 10764283
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 10764283

Brand: Borer Chemie™ D53PLUS.1000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For the pre-cleaning and disinfection of medical instruments, including rigid and flexible endoscopes. Borer Chemie™ deconex™ 53 PLUS Disinfectant is suited for mild treatment and serves as a measure of staff protection.

  • Aldehyde free
  • No active ingredients that contain chloride salts
  • Broad activity spectrum including tuberculocidal effect
  • Active against enveloped viruses and also Adenovirus and Polyoma SV40
  • Particularly gentle on materials
  • Combined cleaning and disinfection effect
  • Suitable for use in ultrasonic bath
  • Certified by the VAH (German Association for Applied Hygiene)

Specifications

pH ∼9
Appearance Clear Green
Certifications/Compliance VAH,CE, 93/42/EEC
Container Type Bottle
Volume (Metric) 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.