Learn More
OHAUS™ Defender™ 5000 W Series Base
Shop All Ohaus Products
Click to view available options
Capacity:
15.00 kg
15.00 kg, 30.00 kg
150.00 kg
150.00 kg, 300.00 kg
3.00 kg, 6.00 kg
30.00 kg
30.00 kg, 60.00 kg
300.00 kg, 600.00 kg
6.00 kg, 15.00 kg
60.00 kg
60.00 kg, 150.00 kg
Model:
D150WQDV
D150WQDX
D150WQV
D150WQX
D150WTDV
D15WQDL
D15WQDR
D15WQL
D15WQR
D300WQDV
D300WTDV
D30WQDL
D30WQL
D600WTDV
D60WQDL
D60WQDV
D60WQDX
D60WQL
D60WQV
D60WQX
D6WQDR
Unit Size:
1 unit
Beschreibung
- The W series features a stainless steel platform, frame and IP67 load cell to withstand harsh industrial environments.
- The Defender 5000 bases are equipped with an EasyConnect cable, which allows instant configuration with Defender 5000 (TD52) indicators, speeding up installation.
- Defender 5000 bases are NTEP-certified and Measurement Canada approved.
- These bases also carry OIML and EC Type approvals, which make them ideal for legal-for-trade use.
Spezifikation
Spezifikation
| Construction | 304-grade stainless steel platform, stainless steel frame, IP67 load cell, non-slip adjustable rubber feet, level indicator |
| Capacity | 150.00 kg, 300.00 kg |
| Readability | 10.000000 g, 20.000000 g |
| Model | D300WQDV |
| Protection Class | IP67 |
| Temperature (Metric) Operating | -10°C to +40°C |
| Dimensions (L x W x H) | 633 x 610 x 130 mm |
| Weight (Metric) | 31.000 kg |