missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Mouse anti-Human, Mouse, Rat, Clone: UMAB166, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies UM570107

This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western BlotSpecifications
Desmin | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P17661, P31001, P48675 | |
Mouse | |
Protein A/G | |
RUO | |
13346, 1674, 64362 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
UMAB166 | |
Unconjugated | |
DES | |
CMD1I; CSM1; CSM2; DES; Desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin | |
Full length human recombiant protein of human DES(NP_001918) produced in E.coli. | |
30 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Desmin Mouse anti-Human, Mouse, Rat, Clone: UMAB166, Origene
Spot an opportunity for improvement?Share a Content Correction