Learn More
Dexamethasone 9,11-epoxide, MedChemExpress
Product Code. 16489056
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Product Code. 16489056
Brand: Medchem Express HYN0348
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Specifications
Chemical Name or Material Dexamethasone 9,11-epoxide
CAS 24916-90-3
Color White
Purity Grade Notes Research
Molecular Formula C22H28O5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O
Molecular Weight (g/mol) 372.45
Formula Weight 372.45
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation