Learn More
Di-tert-butyl phosphite, 95%
Description
Di-tert-butyl phosphite is used as a solvent, as an antioxidant, and as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.995 g/mL |
| Boiling Point | 66°C to 68°C (0.5 mmHg) |
| Refractive Index | 1.42 |
| Quantity | 5 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in organic solvents |
| Formula Weight | 194.21 |
| Percent Purity | 95% |
| Physical Form | Liquid |
| Chemical Name or Material | Di-tert-butyl phosphite |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.