missing translation for 'onlineSavingsMsg'
Learn More
Diethyl benzylidenemalonate, 98%, Thermo Scientific Chemicals Product Code.: 11310114

Diethyl benzylidenemalonate, 98%, Thermo Scientific Chemicals

Product Code. 11310114
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11310114

Brand: Thermo Scientific Alfa Aesar L00561.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethyl benzylidenemalonate is used as a pharmaceutical intermediate. It is also involved in the Conjugate addition of cyanide ion to arylidenemalonic esters provides a useful route to arylsuccinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylidenemalonate is used as a pharmaceutical intermediate. It is also involved in the Conjugate addition of cyanide ion to arylidenemalonic esters provides a useful route to arylsuccinic acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5292-53-5
Molecular Formula C14H16O4
Molecular Weight (g/mol) 248.28
MDL Number MFCD00009149
InChI Key VUWPIBNKJSEYIN-UHFFFAOYSA-N
Synonym diethyl benzylidenemalonate, diethyl benzalmalonate, diethyl 2-benzylidenemalonate, ethyl benzylidenemalonate, diethylbenzalmalonate, diethyl phenylmethylene malonate, malonic acid, benzylidene-, diethyl ester, diethylmalonate, benzal, benzylidenemalonic acid diethyl ester, propanedioic acid, phenylmethylene-, diethyl esterShow MoreShow Less
PubChem CID 94751
IUPAC Name diethyl 2-benzylidenepropanedioate
SMILES CCOC(=O)C(=CC1=CC=CC=C1)C(=O)OCC

Specifications

Melting Point 28°C to 31°C
Density 1.107
Boiling Point 215°C to 217°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 25 g
Beilstein 528894
Solubility Information Insoluble in water.
Formula Weight 248.28
Percent Purity 98%
Chemical Name or Material Diethyl benzylidenemalonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250998 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL00561.14,L00561.22
Lot Number10242936 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog NumberL00561.14,L00561.22
Lot Number10153157 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00561.14,L00561.22
Lot Number10231502 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL00561.14,L00561.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl benzylidenemalonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.