missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cinnamylphosphonate, 98%, Thermo Scientific™

Product Code. 11494567
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17316-55-1
Molecular Formula C13H19O3P
Molecular Weight (g/mol) 254.27
MDL Number MFCD00798606
InChI Key AWKOMEXJXBINBQ-UHFFFAOYSA-N
Synonym diethyl cinnamylphosphonate, diethyl trans-cinnamylphosphonate, 3-phenyl-allyl-phosphonic acid diethyl ester, 2e-3-phenylprop-2-enyl diethoxyphosphino-1-one, diethyl 2e-3-phenylprop-2-en-1-ylphosphonate, diethyl 3-phenylprop-2-en-1-yl phosphonate, diethyl trans-cinnamyl phosphonate, e-cinnamylphosphonic acid diethyl ester, diethyl trans-3-phenyl-2-propenephosphonate
PubChem CID 5914782
IUPAC Name [(E)-3-diethoxyphosphorylprop-1-enyl]benzene
SMILES CCOP(=O)(CC=CC1=CC=CC=C1)OCC

Specifications

Quantity 5 g
Formula Weight 254.27
Percent Purity 98%
Chemical Name or Material Diethyl cinnamylphosphonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.