Learn More
Diethyl sec-butylmalonate, 96%, Thermo Scientific™
Product Code. 11358705
Click to view available options
Quantity:
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83-27-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00015194
InChI Key MIIZSUOEOUHAIZ-UHFFFAOYSA-N
Synonym diethyl sec-butylmalonate, diethyl 2-sec-butyl malonate, sec-butylmalonic acid diethyl ester, diethyl 1-methylpropyl malonate, diethyl 1-methylpropylmalonate, sec-butylmalonic acid, diethyl ester, 1,3-diethyl 2-sec-butyl propanedioate, diethyl s-butylmalonate, diethylsec-butylmalonate, acmc-209prt
PubChem CID 78940
IUPAC Name diethyl 2-butan-2-ylpropanedioate
SMILES CCC(C)C(C(=O)OCC)C(=O)OCC
Specifications
Density 0.98
Boiling Point 110°C to 114°C (18 mmHg)
Quantity 5 g
Beilstein 1709561
Formula Weight 216.28
Percent Purity 96%
Chemical Name or Material Diethyl sec-butylmalonate