missing translation for 'onlineSavingsMsg'
Learn More

Diethylether, >99.5%, Stabilized, Multicompendial, Chem-Lab

Product Code. 13208213
Click to view available options
Quantity:
1 L
Packaging:
GVB
This item is not returnable. View return policy
This item is not returnable. View return policy

Very pure diethylether stabilized with 5-7ppm BHT for laboratory use. Meets DAB, Ph. Eur., BP specifications.

Specifications

CAS 60-29-7
Density 0.71g/mL
Quantity 1 L
Packaging GVB
Molecular Formula C4H10O
MDL Number MFCD00011646
UN Number UN1155
Water <0.2%
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12g/mol
Percent Purity >99.5%
Chemical Name or Material Diethylether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylether v.p.
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Diethylether
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'