missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphinic chloride, Thermo Scientific™

Product Code. 15483747
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15483747

Brand: Thermo Scientific Alfa Aesar H27681.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dimethylphosphinic chloride was used in preparation of dimethyl-ethynylphosphine oxide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-37-4
Molecular Formula C4H10ClOP
Molecular Weight (g/mol) 140.55
MDL Number MFCD00013903
InChI Key KGENPKAWPRUNIG-UHFFFAOYSA-N
Synonym diethylphosphinic chloride, diethylphosphorylchloride, diethylphosphoryl chloride, phosphinic chloride,diethyl, diethylphosphinoyl chloride, acmc-20al83, diethylchlorophosphine oxide, phosphinic chloride, diethyl, 1-chloro ethyl phosphoryl ethane
PubChem CID 5195381
IUPAC Name 1-[chloro(ethyl)phosphoryl]ethane
SMILES CCP(Cl)(=O)CC

Specifications

Refractive Index 1.469
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 140.55
Chemical Name or Material Diethylphosphinic chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.