missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium

For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles

92.00 € - 1520.00 €

Specifications

Certifications/Compliance AOAC, BAM, BS12, COMPF, EP, USDA, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
View More Specs

Products 3
Product Code Brand Quantity Packaging Type Media Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Media Type Price Quantity & Availability  
11782834
View Documents
BD
225650
500 g Bottle Fluid Thioglycollate Medium
92.00 €
500g
In Stock
to see stock.
Edge
Add to basket
 
11742834
View Documents
BD
225620
2000 g Pail Fluid Thioglycollate Medium
515.00 €
2kg
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
11762834
View Documents
BD
225630
10,000 g Pail Fluid Thioglycollate Medium
1520.00 €
10kg
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

  • Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L

Certifications

AOAC, BAM, BS12, COMPF, EP, USDA, USP

Specifications

Specifications

AOAC, BAM, BS12, COMPF, EP, USDA, USP
Dehydrated
Microbiology Media
Used for sterility testing
2°C to 8°C
Fluid Thioglycollate Medium
Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1825 Days

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.