missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Nitrate Broth

Product Code. 11792854
Click to view available options
:
500g
This item is not returnable. View return policy

Product Code. 11792854

Brand: BD 226810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aids in the identification of aerobic and facultative anaerobic, gram-negative microorganisms via the nitrate reduction test. BD™ Difco™ Nitrate Broth is a basal medium containing potassium nitrate. Microorganisms are inoculated into the medium and then incubated.

Microorganisms may be differentiated according to their metabolism of certain substrates. The production of gas from nitrate is an important differential test for glucose-nonfermenting gram-negative bacilli. The end product of reduction depends upon the bacterial species.

  • Nitrate Broth is a basal medium containing potassium nitrate.
  • The microorganism under evaluation is inoculated into the medium and after incubation, nitrate reduction may be determined.
  • Identifies: Acinetobacter calcoaceticus 19606, Enterobacter aerogenes 13048, Escherichia coli 25922, Pseudomonas aeruginosa 27853
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Nitrate Broth
Certifications/Compliance AOAC, BAM, COMPF, ISO, SMD, USDA
Color Light to medium tan
Form Dehydrated (free-flowing, homogenous)
Product Type Microbiology Media
Formulation Beef Extract 3.0g/L; Peptone 4.0g/L; Proteose Peptone (No. 3) 1.0g/L; Potassium Nitrate 1.0g/L
For Use With (Application) For identification of aerobic and facultative anaerobic gram negative microorganisms by means of the nitrate reduction test
Quantity 500 g
compliance-icons
Product Identifier
  • nitrate broth
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : potassium nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.