Learn More
Macherey-Nagel™ Dimethylamino Phases Polygosil™
Product Code. 10328251 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
10 μm
Unit Size:
100g
10g
Product Code. 10328251
Brand: Macherey-Nagel™ 711430.10
Please to purchase this item. Need a web account? Register with us today!
Specifications
Particle Size 10 μm
Pore Size 60 Å
Technique Liquid Chromatography
Quantity 10 g
Type Polygosil Dimethylamino Phases